missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUEDC2 Antibody (08), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06373CL1
This item is not returnable.
View return policy
Description
CUEDC2 Monoclonal antibody specifically detects CUEDC2 in Human samples. It is validated for Flow Cytometry
Specifications
| CUEDC2 | |
| Monoclonal | |
| CoraFluor 1 | |
| bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CUEDC2 (rh CUEDC2; NP_076945.2; aa1-287). | |
| 0.1 mL | |
| Signal Transduction | |
| 79004 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 08 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction