missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CUEDC2 Monoclonal specifically detects CUEDC2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | CUEDC2 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 3 |
| Conjugate | DyLight 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human CUEDC2. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?