missing translation for 'onlineSavingsMsg'
Learn More

CUDC-101, MedChemExpress

Product Code. 16483718
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483718 10 mg 10mg
16466457 5 mg 5mg
16440107 50 mg 50mg
16414796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483718 Supplier Medchem Express Supplier No. HY10223
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CUDC-101 is a potent inhibitor of HDAC, EGFR, and HER2 with IC50s of 4.4, 2.4, and 15.7 nM, respectively.

CUDC-101 is a potent inhibitor of HDAC, EGFR, and HER2 with IC50s of 4.4, 2.4, and 15.7 nM, respectively.

Specifications

Chemical Name or Material CUDC-101
CAS 1012054-59-9
Color White
Purity Grade Notes Research
Molecular Formula C24H26N4O4
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (57.54 mM; Need ultrasonic)
SMILES C#CC1=CC=CC(NC2=NC=NC3=CC(OC)=C(C=C23)OCCCCCCC(NO)=O)=C1
Molecular Weight (g/mol) 434.49
Formula Weight 434.49
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.