Learn More
Sartorius Cubis II Pipette Calibration System
Product Code. 31662882 Shop All Sartorius Products
Change view
Click to view available options
Product Type:
Pipette Calibration System
Unit Size:
Each
Product Code. Product Type Unit Size
31662882 Pipette Calibration System Each
1 options
Product Code. 31662882 Supplier Sartorius Supplier No. MPS116S
Estimated Shipment: 29-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
  • MPS116S is a Cubis™ II-based Single Channel Pipette Calibration System that combines high resolution and large weighing capacity.
  • Together with the option to reduce readability to 10 or 100 g, the instrument is capable of covering up to 95% of the volume ranges for pipettes in the market without sacrificing measurement speed.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pipette Calibration System