missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Cubilin Antibody (SR2571), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-27698-100ul

 View more versions of this product

Product Code. 30041572

  • £307.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cubilin Monoclonal antibody specifically detects Cubilin in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cubilin
Monoclonal
Unconjugated
PBS (pH 7.4), 150mM NaCl, 50% glycerol
Rabbit
Affinity purified
RUO
8029
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG
Western Blot
SR2571
Western Blot 1:500-1:2000
cubilin, cubilin (intrinsic factor-cobalamin receptor), gp280, IFCR, MGA1
A synthesized peptide derived from human Cubilin (Uniprot #: O60494)
100 μL
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.