missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CU070 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524258
This item is not returnable.
View return policy
Description
CU070 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| CU070 | |
| Polyclonal | |
| Unconjugated | |
| FAM207A | |
| 1810008A18Rik; AI303072; C21orf70; Fam207a; family with sequence similarity 207 member A; family with sequence similarity 207, member A; PRED56; Protein FAM207A | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 108707, 85395 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P58468, Q9NSI2 | |
| FAM207A | |
| KLH conjugated synthetic peptide between 146-174 amino acids from the C-terminal region of human CU070. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction