missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CU-CPT22, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Description
CU-CPT22 is a potent protein complex of toll-like receptor 1 and 2 (TLR1/2) inhibitor, and competes with the synthetic triacylated lipoprotein (Pam3CSK4) binding to TLR1/2 with a Ki of 0.41 μM. CU-CPT22 blocks Pam3CSK4-induced TLR1/2 activation with an IC50 of 0.58 μM.
Specifications
Specifications
| Chemical Name or Material | CU-CPT22 |
| CAS | 1416324-85-0 |
| Color | Orange |
| Purity Grade Notes | Research |
| Molecular Formula | C19H22O7 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 125 mg/mL (344.95 mM) |
| SMILES | O=C(C(C=C1O)=CC2=CC(OC)=C(O)C(O)=C2C1=O)OCCCCCC |
| Molecular Weight (g/mol) | 362.37 |
| Formula Weight | 362.37 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction