missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CU CPT 9a

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
CU CPT 9a is a potent TLR8 inhibitor (IC50 = 0.5 nM and Kd = 21 nM). Stabilizes the TLR8 dimer in its resting state, preventing activation. Suppresses TLR8-mediated proinflammatory signaling in various cell lines and human primary cells.
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-4-(7-methoxy-4-quinolinyl)phenol |
| CAS | 2165340-32-7 |
| Quantity | 5 mg |
| Molecular Formula | C17H15NO2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction