missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CU CPT 9a
GREENER_CHOICE

Product Code. 18729006
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18729006

Brand: Tocris Bioscience™ 6479/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent TLR8 inhibitor

CU CPT 9a is a potent TLR8 inhibitor (IC50 = 0.5 nM and Kd = 21 nM). Stabilizes the TLR8 dimer in its resting state, preventing activation. Suppresses TLR8-mediated proinflammatory signaling in various cell lines and human primary cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methyl-4-(7-methoxy-4-quinolinyl)phenol
CAS 2165340-32-7
Quantity 5 mg
Molecular Formula C17H15NO2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.