missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTX-0294885, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Description
CTX-0294885 is a broad spectrum kinase inhibitor that can capture 235 kinases from MDA-MB-231 cells, and can capture all members of the AKT family. CTX-0294885 is a powerful reagent for analysis of kinome signaling networks that can be used for the research of diseases like inflammation, diabetes, and cancer.
Specifications
Specifications
| Chemical Name or Material | CTX-0294885 |
| Physical Form | Solid |
| CAS | 1439934-41-4 |
| Color | Yellowish Green |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H24ClN7O |
| Solubility Information | DMSO : ≥ 100 mg/mL (228.35 mM) |
| SMILES | O=C(NC)C1=CC=CC=C1NC2=NC(NC3=CC=C(N4CCNCC4)C=C3)=NC=C2Cl |
| Molecular Weight (g/mol) | 437.93 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction