missing translation for 'onlineSavingsMsg'
Learn More

CTX-0294885, MedChemExpress

Product Code. 16497026
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497026

Brand: Medchem Express HY15985

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CTX-0294885 is a broad spectrum kinase inhibitor that can capture 235 kinases from MDA-MB-231 cells, and can capture all members of the AKT family. CTX-0294885 is a powerful reagent for analysis of kinome signaling networks that can be used for the research of diseases like inflammation, diabetes, and cancer.

CTX-0294885 is a broad spectrum kinase inhibitor that can capture 235 kinases from MDA-MB-231 cells, and can capture all members of the AKT family. CTX-0294885 is a powerful reagent for analysis of kinome signaling networks that can be used for the research of diseases like inflammation, diabetes, and cancer.

Specifications

Chemical Name or Material CTX-0294885
Physical Form Solid
CAS 1439934-41-4
Color Yellowish Green
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H24ClN7O
Solubility Information DMSO : ≥ 100 mg/mL (228.35 mM)
SMILES O=C(NC)C1=CC=CC=C1NC2=NC(NC3=CC=C(N4CCNCC4)C=C3)=NC=C2Cl
Molecular Weight (g/mol) 437.93
Formula Weight 437.93
Percent Purity 99.46%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.