missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTU2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559441
This item is not returnable.
View return policy
Description
Immunogen sequence: LSPDSFCLGF SEGAACGQSL EERSKTLAEV KPILQATGFP WHVVALEEVF SLPPSVLWCS AQELVGSEGA YKAAVDSFLQ QQH Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 70%.
Cytoplasmic tRNA 2-thiolation protein 2 (CUT2), also named as C16orf84 or NCS2, is a 515 amino acid protein, which belongs to the CUT2 family. C16orf84 localizes in the cytoplasm and Plays a central role in 2-thiolation of mcm5S2U at tRNA wobble positions of tRNA(Lys), tRNA(Glu) and tRNA(Gln). C16orf84 may act by forming a heterodimer with CTU1/ATPBD3 that ligates sulfur from thio-carboxylated URM1 onto the uridine of tRNAs at wobble position.
Specifications
| CTU2 | |
| Polyclonal | |
| Unconjugated | |
| CTU2 | |
| 2310061F22Rik; C16orf84; Ctu2; Cytoplasmic tRNA 2-thiolation protein 2; cytoplasmic tRNA 2-thiolation protein 2 {ECO:0000255; cytosolic thiouridylase subunit 2; cytosolic thiouridylase subunit 2 homolog; cytosolic thiouridylase subunit 2 homolog (S. pombe); HAMAP-Rule:MF_03054}; Ncs2; UPF0432 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 348180 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q2VPK5 | |
| CTU2 | |
| Recombinant protein corresponding to Human CTU2. Recombinant protein control fragment (Product #RP-98583). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction