missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTTNBP2NL Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84648-0.1ML
This item is not returnable.
View return policy
Description
CTTNBP2NL Polyclonal specifically detects CTTNBP2NL in Human samples. It is validated for Western Blot.
Specifications
| CTTNBP2NL | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55917 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| CTTNBP2 N-terminal like, CTTNBP2 N-terminal-like protein, DKFZp547A023, FLJ13278, KIAA1433 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human CTTNBP2NL | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto