Learn More
Invitrogen™ CTTNBP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560789
Description
Immunogen sequence: HDVATDDCKH LLENLNALKI PLRISVGEIE PSNYGSDDLE CENTICALNI RKQTSWDDFS KAVSQALTNH FQAISSDGWW SLEDVTCNNT TDSNIGLSAR SIRSITLGNV PWSVGQSFAQ SPWDFMRKNK AEHITVLLSG PQEGCLSS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
CTTB2 is a protein with six ankyrin repeats and several proline-rich regions. A similar gene in rat interacts with a central regulator of the actin cytoskeleton.
Specifications
| CTTNBP2 | |
| Polyclonal | |
| Unconjugated | |
| CTTNBP2 | |
| 3010022N24Rik; 4732477G22Rik; 6430526E05; 9130022E09Rik; AU040881; brain specific cortactin-binding protein CBP90; C7orf8; Cbp90; cortactin binding protein 2; cortactin-binding protein 2; cortactin-binding protein 2 intron retention form; CORTBP2; Cttnbp2; CTTNBP2 intron retention form; KIAA1758; mKIAA1758; ORF4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83992 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WZ74 | |
| CTTNBP2 | |
| Recombinant protein corresponding to Human CTTNBP2. Recombinant protein control fragment (Product #RP-100929). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.