missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Viral Production Medium

Product Code. 18848745 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
100 L
Unit Size:
10 L
1000mL
20 L
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18848745 6 x 1 L Pack of 6
18838745 1 L 1000mL
18858745 10 L 10 L
18868745 20 L 20 L
31662580 100 L -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18848745 Supplier Gibco™ Supplier No. A5144002
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco CTS Viral Production Medium is a chemically defined, serum-free, protein-free, animal-origin-free, and phenol-red-free medium that is part of the CTS AAV-MAX Helper-Free AAV Production System.

Gibco CTS Viral Production Medium is a chemically defined, serum-free, protein-free, animal-origin-free, and phenol-red-free medium that is part of the CTS AAV-MAX Helper-Free AAV Production System. CTS Viral Production Medium is specifically developed for growth and transfection of CTS Viral Production Cells 2.0 (suspension-adapted HEK293F-derived cells) for adeno-associated virus (AAV) production.

Features of CTS Viral Production Medium include:

  • Supports growth and transfection of CTS Viral Production Cells 2.0 from shake flask to bioreactor scales
  • Enables production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
  • Formulated with GlutaMAX Supplement, so does not require further supplementation. With the exception of containing GlutaMAX Supplement, the formulation of this product is identical to research-grade Viral Production Medium.
  • Manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and follows USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. Drug Master File and Regulatory Support File available upon request

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade (Cat. No. A51217) and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0 (A48400), CTS AAV-MAX Transfection Kit (A5427701), CTS Viral-Plex Complexation Buffer (A5145401), and Viral Production Medium, AGT (A5147101).

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Viral Production Cells 2.0
Cell Type HEK293
Classification Animal Origin-free
Form Liquid
Product Type AAV Production Medium
Serum Level Serum-free
Sterility Sterile-filtered
With Additives GlutaMAX
For Use With (Application) AAV Production
Quantity 6 x 1 L
Shipping Condition Ambient
Format Bottle
Content And Storage 6 x 1 L bottles. Store at 2°C to 8°C and protect from light.
Culture Type Suspension Cell Culture
Expression System Mammalian
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.