missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PureQuant™ Th17 Assay

Product Code. 17116368 Shop All Applied Biosystems Products
Click to view available options
Quantity:
28 reactions
Unit Size:
28 reactions
This item is not returnable. View return policy

Product Code. 17116368

Brand: Applied Biosystems™ A43676

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applied Biosystems™ PureQuant™ assays are a family of assays that accurately identify and quantify specific immune cell types in a mixed population of cells.

Assays are TaqMan qPCR-based and measure genomic DNA methylation at specific sites that serve as a unique identifier. The assays include standards and controls and each 96-well plate can accommodate up to seven test samples. Results are provided as a percentage of target cell type out of total input cells using a free analysis tool.

PureQuant™ Th17 Assay utilizes a specific demethylated site in the pro-inflammatory cytokine interleukin 17A (IL17). This cytokine is exclusively produced by T helper 17 cells.

Benefits include:

  • Proven performance―core technology has been used for identification and quantification of immune cell types for over a decade
  • Easily standardized―qPCR method enables consistent results across multiple users, instruments, and sites
  • Facilitates regulatory filings―regulatory support file provided upon request; established performance specifications following ICHQ2(R1) guidelines
  • Scheduling flexibility―assay can be performed using fresh or frozen cells/gDNA

Assays include all reagents, standards, and controls required to run the assay. Specific components include:

  • Calibrator: Plasmid-based calibrator harbors the genomic target regions in equimolar ratios and includes GAPDH. Subjected to bisulfite conversion followed by copy number estimation via qPCR standard curve. Allows estimation of assay-specific bisulfite efficiency factor: the ratio between TpG/GAPDH copy numbers measured from the same plasmid.
  • Reference: checks assay performance. Consists of genomic DNA extracted from human blood of pooled healthy donors. The reference values represent the expected range of the target cell type. For example, CD8+ cells are generally more abundant than Treg or Th17 cells.
  • Standards: determine the unknown copy number. Consist of six dilutions spanning 4-log linear dynamic range (30-31250 copies). Absolute copy numbers can reliably be determined using the slope and intercept of the respective standard curves.

This product is manufactured at a site with an ISO9001-certified quality management system.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage
  • Dynabeads SILANE Genomic DNA Kit, store at 2°C to 8°CC
  • 1 x 2 mL PureQuant qPCR Master Mix, store at 2°C to 8°CC
  • 4 x ABS
  • THFA, store at room temperature
  • PureQuant Th17 Module, store at -15 to -25°C (CpG & TpG TaqMan primers and probes, Reference DNA, Lambda DNA, Standard DNA)
  • PureQuant Hot Start PCR
  • PureQuant Pre-amplification primer
Detection Method Primer-Probe Detection
For Use With (Application) Cells, DNA
Quantity 28 reactions
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.