missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ OpTmizer™ T-Cell Expansion SFM, no phenol red

Product Code. 30284162 Shop All Gibco Products
Change view
Click to view available options
Quantity:
5 L
10 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30284162 10 L Each
30283668 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30284162 Supplier Gibco™ Supplier No. A3705007
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco™ CTS OpTmizer T-Cell Expansion SFM supports the growth and expansion of human T lymphocytes.

Gibco™ CTS OpTmizer T-Cell Expansion SFM supports the growth and expansion of human T lymphocytes. It is a complete serum-free, xeno-free medium consisting of CTS OpTmizer T-Cell Expansion Basal Medium and CTS OpTmizer T-Cell Expansion Supplement, which are mixed together prior to use.

CTS OpTmizer T-Cell Expansion SFM in 5 L and 10 L bag format is compatible with closed-system processing for media preparation. 5 L and 10 L volumes support scaling of cell therapy manufacturing workflows. The updated bag format allows for easier integration with standard cell therapy equipment, including CTS devices (Xenon, Rotea, and DynaCellect).

Features of CTS OpTmizer T-Cell Expansion SFM include:

  • Both kit components (medium and supplement) in Aegis5-14 film bags
  • Weldable C-Flex and PVC lines
  • Supports high-density T-cell culture (>4 x 106 CD3+ T cells/mL)
  • Supports T-cell activation using Dynabeads magnetic beads, soluble antibodies, and stimulatory antibody-presenting cell protocols
  • Similar phenotype, function (e.g., cytokine secretion profile), and viability to T cells cultured with conventional human AB serum–supplemented medium
  • Supports a T-cell phenotype similar to human serum–supplemented medium
  • Demonstrates enhanced efficacy and persistence of CART-19 cells when grown in medium supplemented with CTS Immune Cell Serum Replacement (ICSR)

Benefits include:

  • Prepare complete medium in a closed system with weldable lines
  • Reduces open steps and contamination risks
  • Easy integration with CTS automated instrumentation

Demonstrated functional performance
Cells grown in CTS OpTmizer T-Cell Expansion SFM with or without human serum demonstrate good cytokine production profiles in polyclonally activated T cells. CTS OpTmizer T-Cell Expansion SFM maintains the desired the CD62L+ central memory T-cell phenotype.

GMP manufactured
The primary manufacturing site for CTS OpTmizer T-Cell Expansion SFM is FDA-registered and has an ISO13485–certified quality management system. The methods and controls used for the manufacturing are in conformity with current Good Manufacturing Practices for medical devices, 21 CFR Part 820, of the regulation.

Versatility
CTS OpTmizer T-Cell Expansion SFM has been widely adopted in clinical T-cell research applications including the culture of CAR T-cells. It works with multiple culture formats: culture flasks, culture bags, and automated and scalable bioreactors. CTS OpTmizer T-Cell Expansion SFM was designed to support expansion of cultured human T-cells without additional serum supplementation. Increased performance is generally observed when supplemented with CTS Immune Cell Serum Replacement.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells
Classification Xeno-free
Form Liquid
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives None
Without Additives No Phenol Red
Product Line OpTmizer
Quantity 10 L
Shipping Condition Room Temperature
Content And Storage • 1 x 10,000 mL bag CTS OpTmizer Expansion Basal Medium, store at 2°C to 8°C, protect from light
• 1 x 260 mL CTS OpTmizer Expansion Supplement in 500 mL bag, store at 2°C to 8°C, protect from light
Culture Environment Bags, Bioreactors
Culture Type Static Culture
Show More Show Less
Are the 5L and 10L sizes/configurations of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107) compatible with CTS devices?

Yes. The 5L and 10L BPCs (bioprocess containers) are weldable to Xenon, Rotea, and DynaCellect consumables.

With the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107), how much supplement should be added to the basal medium?

The 5L and 10L kits are designed so that the entire volume of the supplement BPC (bioprocess container) is added to the basal medium BPC.

With the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107), how is the supplement added to the basal medium?

The supplement BPC (bioprocess container) has 24 in. of sterile weldable 3/32 in. ID, 5/32 in. OD DHEP-free PVC tubing. With fill volumes of 130 mL and 260 mL for the 5L and 10L kits respectively, the supplement BPC can be directly welded to the basal medium BPC.

What are the differences between the BPCs (bioprocess containers) in the 1L sizes of CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM vs. the 5L and 10L sizes?

In the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM, both the basal medium and supplement components are in BPCs (bioprocess containers) composed of Aegis 5-14 film. The basal medium BPC has PVC and C-Flex lines whereas the supplement BPC has a PVC line.

What is the stability of the CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM in the new 5L and 10L sizes?

The 5L and 10L sizes of both CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107) have 12-month expiry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.