missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ KnockOut™ SR XenoFree Kit

Product Code. 10195684
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10195684

Brand: Gibco™ A1099201

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CTS™ KnockOut™ SR XenoFree Kit

GIBCO™ KnockOut™ SR XenoFree CTS™ enables the growth and expansion of human embryonic stem cells (hESC) and human induced pluripotent stem cells (hiPSC) in a cell culture medium containing only human-derived or human recombinant proteins, which facilitates the transition of hESC and hiPSC research from the bench to the clinic.

The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.


  • Defined, xenofree medium for hESC⁄iPSC stem cell culture.
  • First-to-market xenofree hESC⁄iPSC product intended for Ex Vivo tissue and cell culture processing applications.


    GIBCO™ KnockOut™ SR XenoFree CTS™ does not contain bovine or other non-human, animal-derived components. Besides pluripotent stem cell culture expansion and maintenance, KnockOut™ SR XenoFree CTS™ can be used for hESC⁄hiPSC cryopreservation, derivation, and differentiation studies.


    GIBCO™ CTS™ products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate.
    The KnockOut™ SR XenoFree CTS™ Kit includes a bottle of KnockOut™ SR XenoFree CTS™ as a frozen liquid in a 500mL or a 100 mL PET bottle and a product manual.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Cell Type Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs) 
    Classification Serum-free, Xeno-free
    Form Frozen
    Product Type Stem Cell Media
    Serum Level Serum-free
    Sterility Sterile
    With Additives HSA, Sodium Bicarbonate
    Without Additives No Glucose, No Glutamine, No HEPES, No Phenol Red
    For Use With (Application) Cell Therapy, Clinical Research, Translational Research
    Manufacturing Quality cGMP for medical devices, 21 CFR Part 820 and ISO 13485
    Product Line CTS, KnockOut
    Quantity 100 mL
    Endotoxin Level ≤ 10 EU/mL
    pH 6 to 8
    Purity or Quality Grade ES Cell-Qualified
    Species Human
    Content And Storage One bottle

    Store at -20°C to -5°C. Protect from light.
    Culture Environment CO2
    Show More Show Less

    For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.