missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ KnockOut™ DMEM/F-12

Product Code. 12023479 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12023479 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 12023479 Supplier Gibco™ Supplier No. A1370801
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

CTS KnockOut DMEM/F-12 is a serum free, low osmolality medium without L-glutamine or HEPES buffer that is optimized for the growth of human embryonic stem cells (ESC) and induced pluripotent stem cells (iPSC).

Gibco CTS KnockOut DMEM/F-12 is a serum free, low osmolality medium without L-glutamine or HEPES buffer that is optimized for the growth of human embryonic stem cells (ESC) and induced pluripotent stem cells (iPSC). It is designed for use with KnockOut SR and KnockOut SR XenoFree CTS supplements.

Consistent results without fetal bovine serum

Designed for consistent performance, CTS KnockOut DMEM/F-12 is part of the KnockOut line of media products that have delivered reliable results for pluripotent stem cell (ESC and iPSC) culture for years. This fetal bovine free medium leads to little or no spontaneous differentiation.

Equivalent morphology

Stem cells cultured in CTS KnockOut DMEM/F-12 media show normal morphology and karyotype when compared to control stem cells. You can also expect equivalent morphology to manually passaged hESCs, even after enzymatic passaging.

Simplified transition into clinical research

The CTS product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications. Gibco CTS products are of high quality and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files (DMF).

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
Classification Animal Origin-free
Form Liquid
Product Type Cell Culture Medium
Sterility Sterile-filtered
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 500 mL
Shipping Condition Ambient
Format Bottle
Content And Storage Store at 2–8°C and protect from light.
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.