missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Immune Cell Serum Replacement (SR)

Product Code. 31904110
Change view
Click to view available options
Quantity:
50 mL
250 mL
500 mL
1 L
Format:
Bag - BioProcess Container (BPC)
Bottle
Unit Size:
1 L
250mL
500mL
50mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format unitSize
31904110 50 mL Bag - BioProcess Container (BPC) 50mL
15433723 50 mL Bottle 50mL
15463723 500 mL Bottle 500mL
17767187 250 mL Bag - BioProcess Container (BPC) 250mL
17777187 1 L Bag - BioProcess Container (BPC) 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31904110 Supplier Gibco™ Supplier No. A4702903
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

CTS Immune Cell Serum Replacement (SR) is a defined xeno-free formulation designed to support in vitro expansion of human immune cells. It is added as a serum substitute to a basal cell culture medium such as CTS OpTmizer T Cell Expansion SFM or CTS AIM V Medium for the expansion of human immune cells such as T cells.

CTS Immune Cell Serum Replacement (SR) is a defined xeno-free formulation designed to support in vitro expansion of human immune cells. It is added as a serum substitute to a basal cell culture medium such as CTS OpTmizer T Cell Expansion SFM or CTS AIM V Medium for the expansion of human immune cells such as T cells, dendritic cells, and macrophages.

Serum replacement
Human serum, a key component in many immune cell culture protocols, is expensive, inherently variable, and requires extensive safety testing prior to use when manufacturing a cGMP-compliant adoptive cell therapy product. CTS Immune Cell SR was developed to provide a serum-free, xeno-free complete medium that would minimize the supply and safety risks associated with human serum.

CTS Immune Cell SR is a defined and robust formulation that can replace supplementation with human serum in several different base media without sacrificing any performance of your cells. It eliminates the time and cost of qualifying human serum, enabling a seamless transition from research to clinical applications.

Immune cell qualified
CTS Immune Cell SR supports equivalent or improved results for multiple immune cell types (T cells, dendritic cells, macrophages), when used at similar concentration as human serum.

Closed workflow compatible
CTS Immune Cell SR is available in bottle and bioprocess container formats. The bioprocess container chamber is constructed from Aegis 5-14 film. Versatile connection options allow for integration into your closed process.

CTS quality
Gibco CTS products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485-certified. Gibco CTS products follow USP 'ancillary materials for cell, gene, and tissue-engineered products' within the responsibilities applicable to a supplier.*

Accompanied by documentation to support your regulatory filing
Specific intended-use statements, full documentation traceability, and access to our Drug Master File (DMF) are available.

*Other aspects of USP will be the responsibility of the end-user to assess. Thermo Fisher Scientific cannot fulfill USP in regards to application and therapy-specific aspects (e.g., residual assessment and removal of the Ancillary Material).

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Human Immune Cells
Classification Xeno-free
Form Frozen
Product Type Serum Replacement
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives None
Without Additives No Phenol Red
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO13485, MDSAP, FDA-registered, 21CFR820
Quantity 50 mL
Shipping Condition Dry Ice
Format Bag - BioProcess Container (BPC)
Content And Storage -20°C to -5°C. Protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.