Learn More
Gibco™ CTS™ HBSS
Description
Gibco Cell Therapy Systems (CTS) Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. This particular formulation lacks calcium, magnesium and phenol red.
Closed systems compatible
CTS HBSS is available in bottle and bioprocess container formats. The bioprocess container chamber is constructed from Aegis 5-14 film and includes sterile, weldable PVC/C-Flex hybrid tubing with a female Luer lock, providing versatile connection options for integrating into a closed system manufacturing process.
CTS quality
The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.
Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001–certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.
Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis and certificates of origin.
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Culture Type | Mammalian and Stem Cell Culture |
| Form | Clear, colorless solution |
| Product Type | Hanks' Balanced Salt Solution |
| Sterility | Sterile-filtered |
| Sterilization Method | 0.2 μm Sterile-filtered |
| Format | Bag - BioProcess Container (BPC) |
| For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |
| Manufacturing Quality | ISO13485, MDSAP, FDA-registered, 21CFR820 |
| Quantity | 2 L |
| Show More |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.