missing translation for 'onlineSavingsMsg'
Learn More

CTS™ DynaMag™ Magnet

Product Code. 30284220 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30284220 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30284220 Supplier Gibco Supplier No. 12102
 Restricted Item
Estimated Shipment: 25-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CTS DynaMag Magnet

The CTS™ DynaMag™ Magnet is suitable for use with commercially available sterile blood/culture bags, tubing, and connectors.

Benefits:
  • Ideal for magnetic isolation in closed, sterile blood bags
  • Scalable volumes: 50 to 330 mL in static separations and >10 L in continuous flow separations following T-cell expansion protocols
  • Residual beads that escape initial magnetic capture are retained on a secondary magnet

    Applications:
  • Positive isolation of CD3+ T-cells for subsequent stimulation/expansion with CTS™ Dynabeads™ CD3/CD28 and removal of CTS Dynabeads CD3/CD28 following T-cell expansion

    The CTS DynaMag Magnet is intended for use with the CTS Dynabeads product portfolio in clinical research such as CTS Dynabeads CD3/CD28 (Cat. No. 40203D).
  • TRUSTED_SUSTAINABILITY

    Specifications

    Product Line CTS, DynaMag
    Research Category Cell Therapy Research
    Content And Storage The product contains: CTS DynaMag Magnet

    Storage Conditions:
    Protect the device from vibration and keep out of direct sunlight.
    For Use With (Equipment) DynaMag™
    Shipping Condition Room Temperature
    Quantity 1 Each

    For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.