missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ DynaCellect™ Cell Isolation Kit

Product Code. 18229032 Shop All Gibco Products
Change view
Click to view available options
Quantity:
5 kits
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18229032 5 kits Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18229032 Supplier Gibco™ Supplier No. A52300
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Gibco CTS™ DynaCellect™ Cell Isolation Kit is specifically designed for the isolation and activation of immune cells in cell processing applications. The design consists of a 1 L isolation chamber, 8 PVC lines for the welding of input and output bags, a cassette, and tube organizer.

The CTS DynaCellect Cell Isolation Kit is a single-use, fit-for-purpose kit designed specifically for use with the CTS DynaCellect Magnetic Separation System, which is an automated instrument for T-cell isolation and activation, as well as magnetic bead removal.

Characteristics of the CTS DynaCellect Cell Isolation Kit include:
• Weldable tube material: 8 x DEHP-free PVC
• Working volume size: 10 mL to 1 L
• Bag chamber size: 1 L

This kit and the CTS DynaCellect Bead Removal Kit enable a range of cell processing applications such as isolation, depletion, and bead removal. They offer flexibility for process development and can optimized to support commercial scale manufacture.

Each kit has input/output sterile weldable tubes for flexible setup configurations. They are compatible with sterile welding and are gamma sterilized.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Isolation Kit
Capacity (Metric) 1 L
Quantity 5 kits

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.