Learn More
Description
Gibco Cell Therapy Systems (CTS) DPBS is the classic Dulbecco's phosphate-buffered saline. This particular formulation lacks calcium, magnesium, and phenol red.
CTS DPBS is manufactured in cGMP, ISO-certified facilities to help ensure the quality and consistency for reproducible results every day. Tests include osmolality, pH, stability, and the absence of bacterial/fungal contamination and endotoxin.
Gibco CTS products are high quality and supplied with harmonized documentation such as certificates of analysis, certificates of origin, and access to authorization letters for our Drug Master Files, as appropriate. The CTS product line enables you to reduce your burden when qualifying reagents during your transition from research to clinical applications.
Closed systems compatible
This CTS DPBS is provided in bioprocess container format with a chamber constructed from Aegis 5-14 film. The sterile, weldable DEPH-free PVC tubing with Luer lock and C-Flex™ tubing with MPC quick connector provide versatile connection options for closed system manufacturing processes.
Specifications
Specifications
| Chemical Name or Material | DPBS |
| Concentration | 1X |
| For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |
| Physical Form | Liquid |
| Quantity | 2 L |
| Solution Type | Dulbecco's Phosphate Buffered Saline |
| Packaging | Bag |
| Sterility | Sterile-filtered |
| Includes | No Calcium, No Magnesium |
| Manufacturing Quality | ISO13485, MDSAP, FDA-registered, 21 CFR820 |
Frequently Asked Questions (FAQs)
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems. Learn more here.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.