missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ DPBS, calcium, magnesium, bag format

Product Code. 17245996
Click to view available options
Quantity:
2 L
Unit Size:
2 L
This item is not returnable. View return policy

Product Code. 17245996

Brand: Gibco™ A4737901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco Cell Therapy Systems (CTS) DPBS is the classical Dulbecco's phosphate buffered saline, which is a balanced salt solution used for a variety of cell culture applications.

Gibco Cell Therapy Systems (CTS) DPBS is the classical Dulbecco's phosphate buffered saline, which is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. This version contains calcium and magnesium but no phenol red.

CTS quality
The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified. Gibco CTS products follow USP 'ancillary materials for cell, gene, and tissue-engineered products' within the responsibilities applicable to a supplier.

Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis, certificates of origin, and convenient access to our drug master file (DMF).

Closed systems compatible
This CTS DPBS (Cat. No. A4737901, 2 L) is provided in bioprocess container format with a chamber constructed from Aegis 5-14 film. The sterile, weldable DEPH-free PVC tubing with Luer lock and C-Flex™ tubing with MPC quick connector provide versatile connection options for closed system manufacturing processes.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Concentration 1X
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Physical Form Liquid
Quantity 2 L
Solution Type Dulbecco's Phosphate Buffered Saline
Packaging BioProcess Container
Sterility Sterile-filtered
Includes Calcium and Magnesium
Manufacturing Quality ISO13485, MDSAP, FDA-registered, 21 CFR820
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.