missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Detachable Dynabeads™ Release Buffer

Product Code. 30284009 Shop All Gibco Products
Change view
Click to view available options
Quantity:
212 mL
400 mL
750 mL
Unit Size:
212mL
400mL
750mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30284009 212 mL 212mL
30282482 400 mL 400mL
30283272 750 mL 750mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30284009 Supplier Gibco™ Supplier No. A5588303
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Gibco™ CTS Detachable Dynabeads Release Buffer is for use with CTS Detachable Dynabeads magnetic beads. When used together, they empower users with a first-of-its-kind cGMP cell selection technology that has an active release mechanism for process development, clinical trial, and commercial manufacturing use.

The Gibco™ CTS Detachable Dynabeads Release Buffer is for use with CTS Detachable Dynabeads magnetic beads. When used together, they empower users with a first-of-its-kind cGMP cell selection technology that has an active release mechanism for process development, clinical trial, and commercial manufacturing use.

Features of the CTS Detachable Dynabeads Release Buffer (when used with CTS Detachable Dynabeads magnetic beads) include:

  • Empowers users with process flexibility by allowing active release of beads from target cells at any time
  • Optimizes target cell purity, yield, activation profile, and desired phenotype thanks to the active release mechanism that gives users more control
  • Provides an automation-friendly solution by working directly with the CTS DynaCellect Magnetic Separation System
  • Enables consistent performance in light of biological variability within processes
  • Is suitable for both autologous and allogeneic cell therapy manufacturing processes

The CTS Detachable Dynabeads Release Buffer has an active ingredient, biotin, that competitively displaces a biotin derivative on the surface of CTS Detachable Dynabeads magnetic beads. This proprietary mechanism allows users to achieve active release of the beads from target cells at any time within their process. And when used on the CTS DynaCellect Magnetic Separation System, these products deliver automation, flexibility, and scalability in cell therapy manufacturing workflows.

The CTS Detachable Dynabeads Release Buffer also includes Drug Master Files (DMFs) to support cross-referencing in regulatory submissions.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Concentration 5 mM
For Use With (Application) Cell Therapy
For Use With (Equipment) CTS DynaCellect Magnetic Separation System
pH 7.0 to 7.4
Physical Form Liquid
Quantity 212 mL
Solution Type Buffer
Grade GMP

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.