missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Compleo™ Single-Use Kit

Product Code. 31662636 Shop All Gibco Products
Change view
Click to view available options
Quantity:
5 Kits
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31662636 5 Kits
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31662636 Supplier Gibco™ Supplier No. A58165
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Gibco CTS Compleo Single-Use Kit is part of the CTS Compleo Fill and Finish System. The Single-Use Kit supports automated, closed-system processing for formulation and fill workflow steps.

The Gibco CTS Compleo Single-Use Kit is part of the CTS Compleo Fill and Finish System. The Single-Use Kit supports automated, closed-system processing for formulation and fill workflow steps.

Features of the CTS Compleo Single-Use Kit include:

  • Gamma-irradiated, individually packaged, sterile kit
  • 11 user-configurable lines that support attachment through sterile welding or luer lock connection
  • 1 L mixing bag supports volumes up to 750 mL for formulation and filling
  • Kit design reduces residual volume by minimizing the wetted surface area and purging the lines with sterile air after fluid transfer

The CTS Compleo Single-Use Kit provides flexibility for process development and readily supports commercial-scale manufacturing. Cryobags, vials, filters, and other accessories can be easily attached to the Single-Use Kit via sterile welding or luer lock connection.

Tubing material: DEHP-free PVC

Tube dimensions: 4 mm OD, 2.4 mm ID

Pump tubing: PharmaPure

Note: Kits are gamma-irradiated and individually packaged.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 L
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Dimensions Tubing: 3/32 in. I.D., 5/32 in. O.D.
For Use With (Equipment) CTS Compleo Fill and Finish System
Quantity 5 Kits
Sterilization Method Gamma-irradiated
Tubing Line 1 11 weldable tubes - up to 4 inputs and 10 outputs
Tubing Material Line 1 PVC
Tubing Style PVC
Certifications/Compliance ISO 13485
Product Type Single-Use Consumable
Sterility Sterile
Working Volume 750 mL
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.