Learn More
Gibco™ B-27™ Supplement, XenoFree
Description
B-27 Supplement XenoFree is based on the classic B-27 Supplement formulation and contains no animal-derived components at the primary component level. Designed with the translational stem cell researcher in mind, B-27 Supplement XenoFree can be used to support induction of human neural stem cells (hNSCs) from pluripotent stem cells (PSCs), expansion of hNSCs, differentiation of hNSCs, and maintenance of mature differentiated neurons in culture.
Quality
B-27 Supplement XenoFree is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.
Formulated xeno-free
The preparation contains no animal-derived components at the primary component level.
Specifications
Specifications
| Content And Storage | Store in freezer (-5°C to -20°C) and protect from light. |
| Cell Type | Neural Cells |
| Classification | Serum-free |
| Concentration | 50 X |
| Culture Type | Mammalian Cell Culture |
| Form | Liquid |
| Product Type | Cell Culture Supplement |
| Serum Level | Serum-Free |
| Sterility | Sterile |
| pH | 6 to 8 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.