missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ AAV-MAX Transfection Kit

Product Code. 31662651
Click to view available options
Quantity:
1 Kit (For 1 L of production)
1 Kit (For 10 L of AAV production)
1 Kit (For 100 L of production)

Product Code. 31662651

Brand: Gibco™ A5427702

Please to purchase this item. Need a web account? Register with us today!

The Gibco CTS AAV-MAX Transfection Kit is an essential component of the CTS AAV-MAX Helper-Free AAV Production System.

The Gibco CTS AAV-MAX Transfection Kit is an essential component of the CTS AAV-MAX Helper-Free AAV Production System. It is used for the transient transfection of CTS Viral Production Cells 2.0. The kit includes a cationic lipid-based transfection reagent, as well as a transfection booster and production enhancer, which are designed to work together to produce adeno-associated virus (AAV) particles from CTS Viral Production Cells 2.0 when cultured in CTS Viral Production Medium.

Features of the CTS AAV-MAX Transfection Kit include:

  • Components work synergistically within the CTS AAV-MAX system to produce high AAV titers greater than 5 x 1010 vg/mL (unpurified)
  • Robust transfection protocols adaptable to large scale transfection
  • Manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and following USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. Drug Master File and Regulatory Support File available upon request

Components of the kit include:

  • CTS AAV-MAX Transfection Reagent — cationic lipid-based transfection reagent for helper virus-free triple transfection of high-density cultures
  • CTS AAV-MAX Booster — chemically defined component that works with Transfection Reagent to achieve high transfection efficiency
  • CTS AAV-MAX Enhancer — chemically defined component that enhances the production of AAV particles

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade (Cat. No. A51217) and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0 (A48400), CTS Viral Production Medium (A5144001), CTS Viral-Plex Complexation Buffer (A5145401), and Viral Production Medium, AGT (A5147101).

TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin-Free
Content And Storage • 1 x 60 mL bottle of CTS AAV-MAX Transfection Reagent
• 1 x 30 mL bottle of CTS AAV-MAX Transfection Booster
• 1 x 100 mL bottle of CTS AAV-MAX Enhancer

Store at 2°C to 8°C. Do not freeze.

Cell Type HEK293
Format Bottle
Transfection Technique Lipid-based Transfection, Transient Transfection
Expression System Mammalian
For Use With (Application) LV Production
Product Type Transfection Kit
Quantity 1 Kit (For 10 L of AAV production)
Reagent Type Cationic Lipid-Based Transfection
Shipping Condition Gel Packs
Manufacturing Quality ISO 13485, FDA-registered, 21 CFR 820
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.