Learn More
Gibco™ CTS™ AAV-MAX Transfection Kit
Description
The Gibco CTS AAV-MAX Transfection Kit is an essential component of the CTS AAV-MAX Helper-Free AAV Production System. It is used for the transient transfection of CTS Viral Production Cells 2.0. The kit includes a cationic lipid-based transfection reagent, as well as a transfection booster and production enhancer, which are designed to work together to produce adeno-associated virus (AAV) particles from CTS Viral Production Cells 2.0 when cultured in CTS Viral Production Medium.
Features of the CTS AAV-MAX Transfection Kit include:
- Components work synergistically within the CTS AAV-MAX system to produce high AAV titers greater than 5 x 1010 vg/mL (unpurified)
- Robust transfection protocols adaptable to large scale transfection
- Manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and following USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. Drug Master File and Regulatory Support File available upon request
Components of the kit include:
- CTS AAV-MAX Transfection Reagent — cationic lipid-based transfection reagent for helper virus-free triple transfection of high-density cultures
- CTS AAV-MAX Booster — chemically defined component that works with Transfection Reagent to achieve high transfection efficiency
- CTS AAV-MAX Enhancer — chemically defined component that enhances the production of AAV particles
The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade (Cat. No. A51217) and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0 (A48400), CTS Viral Production Medium (A5144001), CTS Viral-Plex Complexation Buffer (A5145401), and Viral Production Medium, AGT (A5147101).
Specifications
Specifications
| Content And Storage | • 1 x 6 mL bottle of CTS AAV-MAX Transfection Reagent • 1 x 3 mL bottle of CTS AAV-MAX Transfection Booster • 1 x 10 mL bottle of CTS AAV-MAX Enhancer Store at 2°C to 8°C. Do not freeze. |
| Format | Kit |
| Transfection Technique | Lipid-based Transfection, Transient Transfection |
| Expression System | Mammalian |
| Packaging Type | Bottle |
| Product Line | AAV-MAX |
| Product Type | Transfection kit for AAV production |
| Phenol Red Indicator | No |
| Quantity | 1 Kit (For 1 L of production) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.