missing translation for 'onlineSavingsMsg'
Learn More

CTRP9/C1qTNF9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-46834

 View more versions of this product

Product Code. 18678265

  • £484.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CTRP9/C1qTNF9 Polyclonal antibody specifically detects CTRP9/C1qTNF9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CTRP9/C1qTNF9
Polyclonal
Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
B2RNN3
Rabbit
Immunogen affinity purified
RUO
338872
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2), 40% Glycerol
AQL1, C1q and tumor necrosis factor related protein 9, C1QTNF9AComplement C1q tumor necrosis factor-related protein 9, complement C1q tumor necrosis factor-related protein 9A, CTRP9, MGC48915
This antibody was developed against Recombinant Protein corresponding to amino acids: DEMWLQVTGGERFNGLFADEDDDTTF
0.1 mL
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.