missing translation for 'onlineSavingsMsg'
Learn More

CTRP7 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18411465 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411465 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411465 Supplier Novus Biologicals Supplier No. NBP176672JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CTRP7 Polyclonal antibody specifically detects CTRP7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTRP7
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias C1q and tumor necrosis factor related protein 7, complement-c1q tumor necrosis factor-related protein 7, CTRP7complement C1q tumor necrosis factor-related protein 7, ZACRP7
Host Species Rabbit
Immunogen Antibody was raised against recombinant human CTRP7.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 114905
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.