missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTRP7 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76634F
This item is not returnable.
View return policy
Description
CTRP7 Polyclonal antibody specifically detects CTRP7 in Human, Mouse, Rat samples. It is validated for ELISA, Immunofluorescence
Specifications
| CTRP7 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunofluorescence | |
| FITC | |
| C1q and tumor necrosis factor related protein 7, complement-c1q tumor necrosis factor-related protein 7, CTRP7complement C1q tumor necrosis factor-related protein 7, ZACRP7 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human CTRP7. The immunogen is located within the last 50 amino acids of CTRP7. Amino Acid Squence: YVDTDYLDSISEDDE | |
| 0.1 mL | |
| Chemokines and Cytokines | |
| 114905 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction