missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CTRP7 Polyclonal antibody specifically detects CTRP7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CTRP7 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C1q and tumor necrosis factor related protein 7, complement-c1q tumor necrosis factor-related protein 7, CTRP7complement C1q tumor necrosis factor-related protein 7, ZACRP7 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human CTRP7. The immunogen is located within the first 50 amino acids of CTRP7. Amino Acid Squence: QPRGNQLKGENYSPR |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?