missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTRP6 Polyclonal Antibody

Description
CTRP6 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | CTRP6 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | C1QTNF6 |
| Gene Accession No. | Q6IR41, Q9BXI9 |
| Gene Alias | 2810036M19Rik; C1q and tumor necrosis factor related protein 6; C1qtnf6; Complement C1q tumor necrosis factor-related protein 6; complement-c1q tumor necrosis factor-related protein 6; CTFP6; CTRP6; UNQ581/PRO1151; ZACRP6 |
| Gene Symbols | C1QTNF6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?