missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CTRP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12828312
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12828312 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12828312 Supplier Invitrogen™ Supplier No. PA520148

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Caco-2 cell lysate. PA5-20148 can be used with blocking peptide PEP-0268.

CTRP2, also known as adiponectin C1q and collagen domain-containing protein 2, is a protein that belongs to the C1q/TNF-related protein family. It is expressed in various tissues, including adipose tissue, skeletal muscle, and the heart, and is involved in the regulation of energy metabolism and insulin sensitivity. CTRP2 has been shown to have anti-inflammatory and cardioprotective effects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTRP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene C1QTNF2
Gene Accession No. Q9BXJ5, Q9D8U4
Gene Alias 1810033K05Rik; C1q and tumor necrosis factor related protein 2; C1qtnf2; Complement C1q tumor necrosis factor-related protein 2; complement-c1q tumor necrosis factor-related protein 2; Crtp2; CTRP2; mCTRP2; RGD1561041; UNQ6349/PRO21054; zacrp2
Gene Symbols C1QTNF2
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the amino-terminus of human CTRP2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114898, 69183
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.