missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTRP2 Polyclonal Antibody

Description
A suggested positive control is NIH-3T3 cell lysate. PA5-20147 can be used with blocking peptide PEP-0267.
CTRP2, also known as adiponectin C1q and collagen domain-containing protein 2, is a protein that belongs to the C1q/TNF-related protein family. It is expressed in various tissues, including adipose tissue, skeletal muscle, and the heart, and is involved in the regulation of energy metabolism and insulin sensitivity. CTRP2 has been shown to have anti-inflammatory and cardioprotective effects.
Specifications
Specifications
| Antigen | CTRP2 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | C1QTNF2 |
| Gene Accession No. | Q9BXJ5, Q9D8U4 |
| Gene Alias | 1810033K05Rik; C1q and tumor necrosis factor related protein 2; C1qtnf2; Complement C1q tumor necrosis factor-related protein 2; complement-c1q tumor necrosis factor-related protein 2; Crtp2; CTRP2; mCTRP2; RGD1561041; UNQ6349/PRO21054; zacrp2 |
| Gene Symbols | C1QTNF2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?