missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CTRP2 Polyclonal antibody specifically detects CTRP2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry
Specifications
Specifications
| Antigen | CTRP2 |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C1q and tumor necrosis factor related protein 2, complement-c1q tumor necrosis factor-related protein 2, CTRP2complement C1q tumor necrosis factor-related protein 2, zacrp2 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against recombinant human CTRP2. |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?