missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTRP2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76671-0.025mg
This item is not returnable.
View return policy
Description
CTRP2 Polyclonal specifically detects CTRP2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.
Specifications
| CTRP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| C1q and tumor necrosis factor related protein 2, complement-c1q tumor necrosis factor-related protein 2, CTRP2complement C1q tumor necrosis factor-related protein 2, zacrp2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 114898 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL | |
| NP_114114 | |
| C1QTNF2 | |
| Antibody was raised against recombinant human CTRP2. | |
| 0.025 mg | |
| Primary | |
| These proteins are often highly modified post-translationally and migrate in SDS-PAGE at positions other than their predicted size. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction