Learn More
Invitrogen™ CTRB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113588
Description
Immunogen sequence: FKNPKFSILT VNNDITLLKL ATPARFSQTV SAVCLPSADD DFPAGTLCAT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 93%.
Preferential cleavage: Tyr- -Xaa, Trp- -Xaa, Phe- -Xaa, Leu- -Xaa.
Specifications
| CTRB1 | |
| Polyclonal | |
| Unconjugated | |
| CTRB1 | |
| 2200008D09Rik; AI504462; Chymotrypsin B; Chymotrypsin B chain A; Chymotrypsin B chain B; Chymotrypsin B chain C; chymotrypsin B1; chymotrypsinogen B; chymotrypsinogen B1; Ctrb; Ctrb1; Prt-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1504 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P17538 | |
| CTRB1 | |
| Synthetic peptide directed towards the middle region of human CTRB1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.