missing translation for 'onlineSavingsMsg'
Learn More

CTR9 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Product Code. 18316413 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18316413 25 μg 25µL
18337262 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18316413 Supplier Novus Biologicals Supplier No. NBP31867725UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CTR9 Polyclonal antibody specifically detects CTR9 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTR9
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200
Formulation Whole antiserum
Gene Accession No. Q6PD62
Gene Alias Ctr9, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae), KIAA0155SH2 domain-binding protein 1, p150TSP, RNA polymerase-associated protein CTR9 homolog, SH2 domain binding protein 1 (tetratricopeptide repeat containing), SH2BP1p150, TPR-containing, SH2-binding phosphoprotein, TSBP
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against mouse CTR9 (ctr9, Paf1/RNA polymerase II complex component), using a KLH-conjugated synthetic peptide containing an amino acid sequence from the N-terminal part of the protein (1).
Purification Method Whole Antiserum
Quantity 25 μg
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 9646
Target Species Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.