missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTR9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21264-1-AP-150UL
This item is not returnable.
View return policy
Description
MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.Specifications
| CTR9 | |
| Polyclonal | |
| Unconjugated | |
| CTR9 | |
| CTR9, KIAA0155, p150, p150TSP, SH2 domain binding protein 1, SH2BP1, TSBP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22083, 293184, 9646 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q62018, Q6PD62 | |
| CTR9 | |
| CTR9 Fusion Protein Ag15690 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction