Learn More
Invitrogen™ CTR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522961
Description
This CTR2 antibody is useful for Western blot, where a band is seen at ~71 kDa. The theoretical molecular weight of CTR2 is 15 kDa. The difference between the expected size and the observed size in Western blot is believed to be due to a multimeric form of CTR2.Suggested positive control: MEF whole cell lysate.
Copper is a trace element that plays an essential role for organisms that utilize oxygen during respiration. CTR2 is a membrane protein that is a member of the SLC31A transporter family. Although the function of CTR2 is still poorly understood in mammalian cells, it is expected to be involved in low-affinity copper uptake. It is believed to be confined to lysosomes which stimulate copper delivery to the the cytosol of human cells at high concentrations.
Specifications
| CTR2 | |
| Polyclonal | |
| Unconjugated | |
| SLC31A2 | |
| AI604396; Copper transporter 2; COPT2; CTR2; hCTR2; probable low affinity copper uptake protein 2; RP11-9H12.2; SLC31A2; solute carrier family 31 (copper transporter), member 2; solute carrier family 31 (copper transporters), member 2; solute carrier family 31 member 2; solute carrier family 31, member 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1318, 20530 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.93 mg/mL | |
| PBS with 30% glycerol and 0.1% sodium azide | |
| O15432, Q9CPU9 | |
| SLC31A2 | |
| Synthetic peptide made to an internal portion of human CTR2 (within residues 50-100). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.