missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595745
This item is not returnable.
View return policy
Description
Immunogen sequence: MHFIFSDEAV LLFDFWRVHS PTGMALSVLV VLLLAVLYEG IKVGKAKLLH KTLESLPATN SQQFILGPDQ DSTGSRSTSD NRTRLRWFLC YFGQSLVHVI; Positive Samples: Mouse pancreas; Cellular Location: Membrane, Multi-pass membrane protein.
Copper is a trace element that plays an essential role for organisms that utilize oxygen during respiration. CTR2 is a membrane protein that is a member of the SLC31A transporter family. Although the function of CTR2 is still poorly understood in mammalian cells, it is expected to be involved in low-affinity copper uptake. It is believed to be confined to lysosomes which stimulate copper delivery to the the cytosol of human cells at high concentrations.
Specifications
| CTR2 | |
| Polyclonal | |
| Unconjugated | |
| SLC31A2 | |
| AI604396; Copper transporter 2; COPT2; CTR2; hCTR2; probable low affinity copper uptake protein 2; RP11-9H12.2; SLC31A2; solute carrier family 31 (copper transporter), member 2; solute carrier family 31 (copper transporters), member 2; solute carrier family 31 member 2; solute carrier family 31, member 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20530 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.43 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9CPU9 | |
| SLC31A2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse Slc31a2 (NP_0012774471). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction