missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTR2 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-05199UV
This item is not returnable.
View return policy
Description
CTR2 Polyclonal specifically detects CTR2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| CTR2 | |
| Polyclonal | |
| Western Blot | |
| Copper transporter 2, COPT2SLC13A2, CTR2Solute carrier family 31 member 2, hCTR2probable low affinity copper uptake protein 2, solute carrier family 31 (copper transporters), member 2 | |
| Rabbit | |
| 0.1 mL | |
| 1318 | |
| Store at 4°C in the dark. |
| Western Blot | |
| DyLight 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SLC31A2 | |
| Synthetic peptide made to an internal portion of human CTR2 (within residues 50-100). [Swiss-ProtNo. O15432] | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction