missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTPS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553606
This item is not returnable.
View return policy
Description
Immunogen sequence: GVCLGMQLAV IEFARNCLNL KDADSTEFRP NAPVPLVIDM PEHNPGNLGG TMRLGIRRTV FKTENSILRK LYGDVPFIEE RHRHRFEVNP NLIKQFEQND LSFVGQDVDG D Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
The protein encoded by this gene catalyzes the formation of CTP from UTP with the concomitant deamination of glutamine to glutamate. This protein is the rate-limiting enzyme in the synthesis of cytosine nucleotides, which play an important role in various metabolic processes and provide the precursors necessary for the synthesis of RNA and DNA. Cancer cells that exhibit increased cell proliferation also exhibit an increased activity of this encoded protein. Thus, this protein is an attractive target for selective chemotherapy. Alternative splicing results in multiple transcript variants.
Specifications
| CTPS2 | |
| Polyclonal | |
| Unconjugated | |
| Ctps2 | |
| A830031M15Rik; AI326475; CTP synthase 2; CTP synthase II; CTP synthetase 2; CTP synthetase homolog; CTP synthetase type 2; CTPS2; CTPsH; cytidine 5'-triphosphate synthase 2; cytidine 5'-triphosphate synthetase 2; cytidine triphosphate synthase II; UTP-ammonia ligase 2; UTP--ammonia ligase 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56474, 619580 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.30 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5U2N0, Q9NRF8 | |
| Ctps2 | |
| Recombinant protein corresponding to Human CTPS2. Recombinant protein control fragment (Product #RP-91884). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction