missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTPS Recombinant Rabbit Monoclonal Antibody (23GB1685), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CTPS |
|---|---|
| Clone | 23GB1685 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089274
|
Invitrogen™
MA553734 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089912
|
Invitrogen™
MA553735 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CTPS Recombinant Monoclonal antibody specifically detects CTPS in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| CTPS | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P17812, P70698 | |
| 1503, 313560, 51797 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1685 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| cb1040; CTP synthase; CTP synthase 1; CTP synthase 1a; CTP synthase a; CTP synthetase 1; CTPS; ctps1; ctps1a; ctpsa; cytidine 5-prime triphosphate synthetase; cytidine 5'-triphosphate synthase; cytidine 5'-triphosphate synthetase; IMD24; UTP--ammonia ligase 1; wu:fb49e03; wu:fe17b03 | |
| A synthesized peptide derived from human CTP synthase/CTPS (250-300AA). | |
| Primary | |
| -20°C | |
| CTPS1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title