missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CTP Solution, Tris buffered

Product Code. 15245116 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.25 mL
Unit Size:
25000nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15245116 0.25 mL 25000nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15245116 Supplier Thermo Scientific™ Supplier No. R1451
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific CTP (cytidine 5'-triphosphate), Tris buffered is an extremely stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with Tris base.

Thermo Scientific CTP (cytidine 5'-triphosphate), Tris buffered is an extremely stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with Tris base. The nucleotide has greater than 99% purity and is free of nuclease activities and functionally tested in in vitro transcription.

Features
• Purity confirmed by HPLC
• Free of trace contaminating nucleotides, endo-, exodeoxyribonuclease and ribonuclease activities
• Higher in vitro transcription yields using Tris buffered NTPs in comparison to NTPs titrated with NaOH
• Stable for 3 years at -20°C and after multiple freeze-thaw cycles

Applications
• in vitro transcription
• RNA amplification
• siRNA synthesis
• aRNA synthesis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 100 mM CTP, Tris buffered (0.25 mL)

Store at –15 to –25°C.
Shipping Condition Dry Ice
Concentration 100 mM
Label or Dye Unlabeled
Quantity 0.25 mL
For Use With (Application) in vitro Transcription, RNA Amplification
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.