missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTP synthase Antibody (S01-9E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19612-50ul
This item is not returnable.
View return policy
Description
CTP synthase Monoclonal antibody specifically detects CTP synthase in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CTP synthase | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S01-9E5 | |
| Unconjugated | |
| CTP synthase, CTP synthase 1, CTP synthetase 1, cytidine 5-prime triphosphate synthetase, cytidine 5'-triphosphate synthetase, EC 6.3.4.2, UTP--ammonia ligase 1 | |
| A synthetic peptide of human CTP synthase (Uniprot # P17812) | |
| 50 μg | |
| Endocrinology, Signal Transduction | |
| 1503 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction