missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTNS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13085-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody was raised against the N-terminal region of human cystinosin. Two isoforms of CTNS exist due to alternative splicing events and both of them can be detected through this antibody.
This gene encodes a seven-transmembrane domain protein that functions to transport cystine out of lysosomes. Its activity is driven by the H+ electrochemical gradient of the lysosomal membrane. Mutations in this gene cause cystinosis, a lysosomal storage disorder. Alternative splicing results in multiple transcript variants.Specifications
| CTNS | |
| Polyclonal | |
| Unconjugated | |
| CTNS | |
| CTNS, CTNS LSB, Cystinosin, cystinosis, nephropathic, PQLC4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1497, 287478, 83429 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60931, P57757 | |
| Ctns | |
| CTNS Fusion Protein Ag3904 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction